Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Spunbond Polypropylene Coverall

Catalog No. 19046231 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
2X-Large
Color:
Navy
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-046-231 2X-Large Navy
19-821-590 Large White
19-821-591 Medium White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-046-231 Supplier West Chester Supplier No. 3575/XXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spunbound Polypropylene offers lightweight protection against dry particulates, keeping dirt and debris offs the body. Good for use in General labor, pharmaceuticals, cleaning and food processing.

  • Protection against dry particulates
  • For use in General labor, pharmaceuticals, cleaning and food processing

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Spunbond Polypropylene
Color Navy
Disposable Single-use
Waist Style Loose
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper (Full-length)
Seam Type Serged
Sterility Non-sterile
Resistance Dust
Latex Free Yes
No. of Pockets None
Size (Metric) Inseam 96 cm
For Use With (Application) Agriculture, general industry, construction and municipalities
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.