Learn More
West Chester Spunbond Polypropylene Lab Coat
Catalog No. 190881685 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Thickness:
Standard weight
Size:
Large
X-Large
3X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness Size
19-088-1685 Standard weight X-Large
19-088-1684 - Large
19-088-1689 - 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-088-1685 Supplier West Chester Supplier No. 3511/XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Standard weight lab coats offer breathability and durability. Choose open wrists or elastic wrists style.
  • Lightweight protection against dry particulates
  • Made with durable polypropylene material
  • Snap front closure for secure fit and allows easy donning
  • Collared neck for better air circulation
  • Choose open or elastic wrist style
Specifications
Size X-Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
Resistance Dust
Style Full-length
No. of Pockets None
Latex Free Yes
Thickness Standard weight
Autoclavable Not Autoclavable
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.