Learn More
West Chester™ Standard Split Cowhide Leather Welding Gloves
Catalog No. 19046225 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
Catalog No.
19-046-225 Large
1 options
This item is not returnable. View return policy
Catalog No. 19-046-225 Supplier West Chester™ Supplier No. 930
This item is not returnable. View return policy
Split leather gloves provide heat protection for general welding applications. Cotton sock lining absorbs perspiration, and the one piece palm does not have any exposed seams.
  • Reinforced welted seams
  • Cotton sock lining absorbs perspiration
  • Extended gauntlet cuff provides lower arm protection from sparks
  • Gunn pattern
  • Wing thumb
Specifications
Color Gray
Material Leather
Size Large
Glove Cuff Style Gauntlet
Length (English) 13.5 in.
Length (Metric) 34.3 cm