Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ Standard Split Cowhide Leather Palm Gloves

Catalog No. 19821540 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-821-540 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-540 Supplier West Chester™ Supplier No. 100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Striped canvas gloves with a leather palm and fingers provide protection and finger dexterity.

  • Split cowhide leather palm and fingers
  • 5 piece striped canvas back
  • Knit wrist helps keep dirt and debris out of the glove

Specifications

Color Blue, Red
Grade Standard
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type Leather Palm Gloves
Size Large
Glove Cuff Color Natural
Glove Cuff Style Knit
Liner Material Polycotton
Yarn Weight 8 oz.
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.