Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix West Nile Virus Culture Fluid
SDP

Catalog No. 22157583 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Product Type:
West Nile Virus 2
West Nile Virus 956-99 Lineage 2
West Nile Virus NYS-1
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-157-583 West Nile Virus NYS-1
22-157-637 West Nile Virus 956-99 Lineage 2
22-157-639 West Nile Virus 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-157-583 Supplier ZeptoMetrix Supplier No. 0810033CF
Add to Cart
Add to Cart

Virus preparation containing live whole organisms for various research and product development needs

IMPORTANT: Contains live material, so cust must submit an executed ZeptoMetrix MTA before vendor can ship. Also, a USDA APHIS VS 16-6 or 16-6A authorized permit must be obtained and a valid copy of the permit received by ZeptoMetrix prior to shipment of this product. requests.

Specifications

Certifications/Compliance ISO, 13485
Content And Storage -65°C
Form Solution
Media Type Vial
Product Type West Nile Virus NYS-1
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.