Learn More
Description
West Nile Virus Envelope Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | West Nile Virus Envelope |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | West Nile envelope; West Nile Virus; WNV envelope |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to 73-86 of West Nile Virus Envelope glycoprotein. |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
