Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ West Nile Virus NS3 Protein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111984 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111984 Supplier Invitrogen™ Supplier No. PA5111984
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

West Nile Virus NS3 Protein Polyclonal Antibody for Western Blot, ICC/IF

West Nile virus (WNV) is an infectious disease that first appeared in the United States in 1999. Infected mosquitoes spread the virus that causes it. People who get WNV usually have no symptoms or mild symptoms. The symptoms include a fever, headache, body aches, skin rash, and swollen lymph glands. The West Nile Virus (WNV) NS3 protease is absolutely essential (along with viral-encoded cofactor NS2B) for post-translational processing of a viral polypeptide precursor in infected host cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen West Nile Virus NS3 Protein
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias West Nile Virus
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of West Nile virus NS3 protein (West Nile virus (strain NY99-IC)). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.