Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Western Blot Marker Pen, For PVDF And Nitrocellulose, 1 Ea.

Catalog No. NC1069503
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC1069503 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC1069503 Supplier Abcam Supplier No. AB1668581UNIT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Marker pen for annotating blots and molecular weight markers on PVDF or nitrocellulose membranes.

The Western Blot Marker Pen is designed for annotating western blots and marking the positions of molecular weight standard markers on PVDF or nitrocellulose membranes.

  • Constituents: 10% Glycerol and 1% C.I. Acid Blue 83
  • Storage: Store at +4°C
  • Shipping: Blue Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Format Pen
Form Liquid
Membrane Compatibility Nitrocellulose, PVDF
Shipping Condition Blue Ice
Quantity 1 Ea.
For Use With (Application) Western Blot
Storage Requirements Store at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.