Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WFDC2 Recombinant Rabbit Monoclonal Antibody (JF62-09)
GREENER_CHOICE

Catalog No. PIMA532535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532535 Supplier Invitrogen™ Supplier No. MA532535
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This gene encodes a protein that is a member of the WFDC domain family. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. This gene is expressed in pulmonary epithelial cells, and was also found to be expressed in some ovarian cancers. The encoded protein is a small secretory protein, which may be involved in sperm maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WFDC2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone JF62-09
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene WFDC2
Gene Accession No. Q14508, Q9DAU7
Gene Alias 1600023A02Rik; dJ461P17.6; EDDM4; epididymal protein 4; epididymal secretory protein 4; Epididymal secretory protein E4; epididymis-specific, whey-acidic protein type, four-disulfide core; HE4; Major epididymis-specific protein E4; MGC57529; putative protease inhibitor WAP5; RE4; WAP domain containing protein HE4-V4; WAP domain-containing protein HE4; WAP four-disulfide core domain 2; WAP four-disulfide core domain protein 2; WAP5; Wfdc2
Gene Symbols WFDC2
Host Species Rabbit
Immunogen Synthetic peptide within Human HE4 aa 78-114.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10406, 67701
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.