Learn More
Apexbio Technology LLC WH-4-023, 10mg. Cas: 837422-57-8 MFCD: MFCD28138072. Lck/Src inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC B800310
Encompass_Preferred
WH-4-023 is a potent and selective Lck and Src inhibitor with IC50 values of 2 and 6 nM, respectively [1]. It is also an inhibitor of IC50 values of 10, 22 and 60 nM for SIK 1, 2 and 3, respectively [2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5449
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.