Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ WHA200 Auto Dark Helmet

Catalog No. 19156097 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Material: Thermoplastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-156-097 Material: Thermoplastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-156-097 Supplier Pyramex™ Supplier No. WHA200

missing translation for 'validMainframeMsg'

Auto darkening welding helmet with sensitivity adjustments

  • Lightweight thermoplastic shell provides total head, face and neck protection and eliminates welder's fatigue
  • Fully adjustable nylon suspension
  • Adjusts from IR shade #9-13 Sensitivity adjustment
  • Battery (2/AAA) and solar powered with 1 sensor
  • Light state is shade #3
  • High speed lens - 1/25000 second
  • Optimally balanced shake down helmet minimizes carbon monoxide build up
  • Ample space for respirators
  • Specifications

    Certifications/Compliance ANSI Z87.1-2003
    Color Black
    For Use With (Application) Welding
    Material Thermoplastic
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.