Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crabtree What Are Food Chains and Webs? by Bobbie Kalman
SCI_ED

Catalog No. S000946 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Format:
Paperback Book
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S000946 Paperback Book
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000946 Supplier Crabtree Supplier No. 9780865058880
Add to Cart
Add to Cart

Educational book about food chains and webs.

  • Book by Bobbie Kalman from Crabtree, part of the Science of Living Things series

Specifications

Grade 2
Format Paperback Book
Product Type Paperback, Food Chains
Languages English
Subject Food Chains, Eco Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.