Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teacher Created Materials What do Living Things Need? by Elizabeth Austen
SCI_ED

Catalog No. S000941 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Format:
Paperback Book
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S000941 Paperback Book
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000941 Supplier Teacher Created Materials Supplier No. AB5310
Add to Cart
Add to Cart

Literature for educational purposes

  • Educational Book

Specifications

Grade 1
Format Paperback Book
Product Type Paperback, Human Needs
Subject Living Organisms
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.