Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ 0.8 μm Pore Size (ME 27) Mixed Cellulose Ester Membranes, White/Black Grid

Catalog No. 09927365 Shop All Cytiva Products
Change view
Click to view available options
Filter Type:
Gridded Filter with Pad
Gridded Filter without Pad
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type
09-927-365 Gridded Filter without Pad
09-927-275 Gridded Filter with Pad
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-927-365 Supplier Cytiva Supplier No. 10408970
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Whatman™ (ME 27) Mixed Cellulose Ester Membranes, White/Black Grid are particularly effective in applications requiring higher flow rates and larger volume filtration. Supplied with clearly defined grid lines at regularly spaced intervals to assist in microbiological colony counting.

  • Material: Mixed Cellulose Ester
  • Material (Membrane): ME 27
  • Pore Size: 0.8 μm
  • Size (Metric) Grid: 3.1 mm
  • Color: White
  • Grid Color: Black
  • Autoclavable: Yes
  • Sterility: Sterile
  • Thickness (Metric): 140 μm
  • Diameter (Metric): 47 mm
  • Flow Rate: 2.8 sec./100 mL/12.5cm² (at Δp = 0.9 bar)
  • Gas Flow Rate: 11.6 sec./100 mL (at Δp = 3 mbar)
  • Bubble Point: 1.3 bar
  • For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Bubble Point 1.3 bar
Color White
Filter Type Gridded Filter without Pad
Material MCE
Material (Membrane) ME 27
Pore Size 0.8 μm
Sterility Sterile
Flow Rate 2.8 sec./100 mL/12.5 cm2 (at Δp = 0.9 bar)
For Use With (Application) Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Gas Flow Rate 11.6 sec./100 mL (at Δp = 3 mbar)
Grid Color Black
Quantity 100 Pk.
Size (Metric) Grid 3.1 mm
Thickness (English) 0.006 in.
Thickness (Metric) 140 μm
Diameter (English) 1.85 in.
Diameter (Metric) 47 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.