Learn More
Cytiva Whatman™ 903 Proteinsaver Snap-Apart Card
GSA_VA
Catalog No. 09800961 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
4 x 75 to 80μL
Catalog No. Volume (Metric)
09-800-961 4 x 75 to 80μL
1 options
Catalog No. 09-800-961 Supplier Cytiva Supplier No. 10534320
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
903 Sample Collection Card with 4 sample spots.
903 sample collection card with 4 sample spots.
The paper in our 903 Proteinsaver Snap-Apart Card, imprinted with 4 half-inch circles, is enclosed between two pieces of cover stock. Each circle holds 75-80 µl of sample. To use the 903 Proteinsaver Snap-Apart Card, one cover is snapped off, the sample is collected and the remaining cover is folded over the sample. This cover is imprinted with the universal biohazard symbol.
  • Ready-to-use cards and kits save time and costs during sample collection, transport, and archiving.
  • Strict quality control and ISO9001 manufacturing standards ensure high quality, reproducibility, and purity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Material Cellulose
For Use With (Application) Sample collection, transport, and archiving
Includes 4 Sample Spots
Product Line Whatman
Type 903 Proteinsaver Snap-Apart Card
Volume (Metric) 4 x 75 to 80μL