Learn More
Cytiva Whatman™ Antibiotic Assay Paper
Catalog No. 09927813 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Outside:
9 mm
Catalog No. Diameter (Metric) Outside
09-927-813 9 mm
1 options
Catalog No. 09-927-813 Supplier Cytiva Supplier No. 2017009
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
For determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
Antibiotic assay discs
The antibiogram allows rational and selective chemotherapy. The test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated. The size of the inhibition zone is a measure for the effectiveness of the substances.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Antibiotic Assay Paper
For Use With (Application) For research use, in determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method
Diameter (Metric) 9 mm
Diameter (Metric) Outside 9 mm