Learn More
Cytiva Whatman™ AS 300 Series Filtration Manifolds
Catalog No. 09806202 Shop All Cytiva Products
Change view
Click to view available options
No. of Positions:
3
Catalog No. No. of Positions
09-806-202 3
1 options
Catalog No. 09-806-202 Supplier Cytiva Supplier No. 10498761
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Stainless steel filter funnel three-place manifold, for microbiology monitors and analytical funnels
The stainless steel manifold for three or six filtration units is fitted with stainless steel units. The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Manifold Type 3 Stopcocks and lower parts for individual choice of filter units
No. of Positions 3
Port Detail 9 mm I.D.
For Use With (Equipment) Microbiology monitors and analytical funnels