Learn More
Cytiva Whatman™ AS 600 Series Filtration Manifold
Catalog No. 09800960 Shop All Cytiva Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
09-800-960 Stainless Steel
1 options
Catalog No. 09-800-960 Supplier Cytiva Supplier No. 10498762
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Stainless steel filter funnel six-place manifold, for microbiology monitors and analytical funnels
The stainless steel manifold for three or six filtration units is fitted with stainless steel units. The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Manifold Type 6 Stopcocks and lower parts for individual choice of filter units
Port Detail 9 mm I.D.
For Use With (Equipment) Microbiology monitors and analytical funnels