Learn More
Cytiva Whatman™ AS 600 Series Multiple Vacuum Filtration Apparatus
Catalog No. 09927563 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
Includes:
Stainless Steel support screen filter, 6 place manifold
Stainless Steel support screen filter, 6 place manifold, rapid closure clamp
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Includes
09-927-563 100 mL Stainless Steel support screen filter, 6 place manifold, rapid closure clamp
09-927-413 500 mL Stainless Steel support screen filter, 6 place manifold, rapid closure clamp
09-927-310 500 mL Stainless Steel support screen filter, 6 place manifold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-927-563 Supplier Cytiva Supplier No. 10444850
AS610/3 Multiple Vacuum Filtration Apparatus, six-place vacuum filtration system, stainless steel 500 ml, 47/50 mm, support screen, with rapid closure clamp
AS 600 Series
The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Includes Stainless Steel support screen filter, 6 place manifold, rapid closure clamp
Series AS600/5 Series
Type Vacuum Filtration System
Volume (Metric) 100 mL