Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Benchkote™ Plus Surface Protector Sheets PROMO

Catalog No. 12007183AL2 Shop All Cytiva Products
Change view
Click to view available options
Dimensions (L x W):
500 x 600 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
12-007-183AL2 500 x 600 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-007-183AL2 Supplier Cytiva Supplier No. 29822151

missing translation for 'validMainframeMsg'

Prevent many common sources of laboratory contamination, such as those involving hazardous materials on shared workspace benches. Whatman™ Benchkote Plus bench liner can also help with the disposal of hazardous materials.

  • Strong: Benchkote Plus bench liner is strong and tear resistant when wet or dry
  • Ease-of-use: Smooth white surface can be written on with ink or pencil and lies flat
  • Clean: Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Disposable: Benchkote Plus bench paper can be incinerated after use
  • Safe combustion: Polyethylene layer is rapidly consumed when incinerated and does not melt or drip
TRUSTED_SUSTAINABILITY

Specifications

Product Type Surface Protector
Material Polyethylene
Absorbancy 750 mL/m2
Length (English) 19 in.
Length (Metric) 500 mm
Width (English) 23 in.
Width (Metric) 600 mm
Dimensions (L x W) 500 x 600 mm
For Use With (Application) Suitable for saturation with disinfectant
Quantity 50 Sheets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.