Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Black Cyclopore™ Membranes

Catalog No. 09927986 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric):
47 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
09-927-986 47 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-927-986 Supplier Cytiva Supplier No. 70634702

missing translation for 'validMainframeMsg'

For use with epifluorescence microscopy and applications requiring contrasting background.

  • Black polycarbonate membrane
  • Dark membrane reduces background fluorescence
  • Improves sensitivity of test
  • Membrane used to filter test and then used for analysis
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color Black
Filter Type Membrane Filter
Material Polycarbonate
Pore Size 0.2 μm
For Use With (Application) Highly suitable for epifluorescence and other microscopy applications
Hydrophobicity Hydrophilic
Type Filtration Membrane Disc
Diameter (Metric) 47 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.