Learn More
Cytiva Whatman Cyclopore Polycarbonate Membrane
Catalog No. 18400199 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.4 μm
Diameter (Metric):
293 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Diameter (Metric)
18-400-199 - -
18-400-195 - -
18-400-196 0.2 μm 293 mm
18-400-197 0.4 μm 293 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-400-199 Supplier Cytiva Supplier No. 10417450
Add to Cart
Add to Cart
Featuring sharp cut-off and reproducible microfiltration characteristics of track-etched membrane filters
  • Low affinity for stains improves visibility under microscope
  • Low protein binding minimizes sample loss of proteinaceous samples
  • Narrow pore size distribution and high pore density support rapid surface capture of analyte
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • No particle shedding enables clean filtrate
  • Biologically inert
TRUSTED_SUSTAINABILITY
Specifications
Material Polycarbonate
Quantity 100 Pk.