Description
- Prepare 96 samples up to 150μL for plasma and 200μL for serum in less than 10 minutes
- Minimal liquid handling—precipitation and filtration performed in same well
- Utilizes acetonitrile precipitation and vacuum filtration to remove protein contaminants from samples before downstream analysis
- Oleophobic membrane configuration retains acetonitrile/sample mixture during mixing
- Vacuum filtration allows mixture to pass through the membrane while precipitated proteins remain in well
- Filtrate is then ready for HPLC injection or target UV quantitation
Plasma monitoring for pharmaceutical efficacy and new drug development
Specifications
Specifications
| Description | GE’s Whatman UNIFILTER 2 mL, 96 well microplates are used for large sample or reagent volumes, as in biomolecule purification by SPE and organic synthesis in combinatorial chemistry library generation. |
| Array | 8 x 12 |
| Product Type | UNIFILTER Microplate, 96-well, 2 mL, protein precipitation fast flow, filter bottom with long drip d |
| Material | Glass-filled Polypropylene |
| No. of Wells | 96 |
| Well Shape | Filter |
| For Use With (Application) | Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation |
| Includes | Long drip director |