Learn More
Cytiva Whatman™ Grade 91 Wet Strengthened Qualitative Filter Papers - Circles
Shop All Cytiva ProductsDescription
High wet strength due to the addition of chemically stable resin
- Wet strengthened grade
- General purpose creped filter
- For less critical routine analysis
- Used to assay sucrose in cane sugar
Specifications
Specifications
| Description | Grade 91 Qualitative Filter Paper Wet-Strengthened, circle, 110 mm (subdivided into 100) |
| Format | Disc |
| Material | Cellulose |
| Particle Retention | 10 μm |
| Pore Size | 10 μm |
| Filtration Speed | 70 sec. |
| For Use With (Application) | A general purpose creped filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration. |
| Grade | Grade 91 |
| Quantity | 40 x 100 Pk. |
| Thickness (Metric) | 205 μm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.