Learn More
Cytiva Whatman™ Hepa-Cap™ In-Line Venting Filters
Catalog No. 09800977 Shop All Cytiva Products
Change view
Click to view available options
Certifications/Compliance:
HEPA-CAP 36
HEPA-CAP 75
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
09-800-977 HEPA-CAP 36
09-800-980 HEPA-CAP 75
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-800-977 Supplier Cytiva Supplier No. 2609T
Add to Cart
Add to Cart
For use in scientific, research and industrial environments where high retention, dirt-holding capacity and flow rates are required
  • Dual lamination with polyester monofilament
  • Retains 99.97% of all particles ≥ 0.3μm in air
  • Suitable for particulate removal from air and gases
  • Able to be sterilized by autoclaving with steam
  • Allows bidirectional flow
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Certifications/Compliance HEPA-CAP 36
Pore Size 0.3 μm
For Use With (Application) For use in scientific, research and industrial environments where high retention, dirt-holding capacity and flow rates are required
Housing Material Polypropylene
Inlet Style 3/8 in. FNPT
Max. Pressure 4.1 bar (60 psi)
No. per Pack 5 Pieces
Outlet Style 3/8 in. FNPT
Quantity 5/Pk.
Type Inline Venting Filter