Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Litmus Blue Test Paper
GSA_VA

Catalog No. 0987577 Shop All Cytiva Products
Change view
Click to view available options
Format:
Reel
Strip
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
09-875-77 Reel
09-924-251 Strip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-875-77 Supplier Cytiva Supplier No. 2600201A
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Impregnated with a mixture of pH indicators, acid-alkali litmus papers assume a color specific to sample pH upon contact with a sample. Whatman™ Litmus Blue Test Paper provides rapid determination of pH values and has a variety of applications in laboratories and industry.

Mixed-indicator tests designed for fast, accurate, and reliable pH solution testing.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
  • Color change in pH solutions ranging from approximately 5 to 8
  • Economical

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Indicator Blue
pH Range 5 to 8
Format Reel
Quantity 1 Each
Type Test Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.