Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Litmus Blue Test Paper

Catalog No. 09924251 Shop All Cytiva Products
Change view
Click to view available options
Format:
Reel
Strip
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
09-924-251 Strip
09-875-77 Reel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-924-251 Supplier Cytiva Supplier No. 10360300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Impregnated with a mixture of pH indicators, acid-alkali litmus papers assume a color specific to sample pH upon contact with a sample. Whatman™ Litmus Blue Test Paper provides rapid determination of pH values and has a variety of applications in laboratories and industry.

Mixed-indicator tests designed for fast, accurate, and reliable pH solution testing.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
  • Color change in pH solutions ranging from approximately 5 to 8
  • Economical
TRUSTED_SUSTAINABILITY

Specifications

Color Indicator Blue
pH Range 5 to 8
Format Strip
Quantity 100 Pack
Type Test Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.