Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ pH Indicators Reel Dispensers
GSA_VA

Catalog No. 0987570 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
0.5 to 5.5
1 to 11
1 to 14
4 to 7
6.4 to 8
8 to 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
0987570 1 to 14
09-875-71 0.5 to 5.5
09-875-73 6.4 to 8
09-876-16 1 to 11
09-875-72 4 to 7
09-927-820 8 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 0987570 Supplier Cytiva Supplier No. 2600100A
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for rapid determination of pH values

  • Instant pH readings
  • Accurate for wide range of routine pH testing
  • Convenient and portable for field use

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

pH Range 1 to 14
Format Reel
For Use With (Application) Routine pH testing
Type pH Indicator Dispenser
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.