Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Polycap™ TC High-Capacity Capsules
GSA_VA

Catalog No. 05713750 Shop All Cytiva Products
Change view
Click to view available options
Filtration Area:
1900 cm2
440 cm2
930 cm2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filtration Area
05-713-750 440 cm2
05-713-751 930 cm2
05-713-752 1900 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-713-750 Supplier Cytiva Supplier No. 67153682
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Edge
Add to Cart

Developed for safe, efficient filtration of critical biological and tissue culture solutions. Whatman™ Polycap™ TC High-Capacity Capsules include PES membranes for excellent flow rate and protein absorption characteristics.

Contain two layers of polyethersulfone membrane — inherently hydrophilic, low in extractables, excellent flowrates. Low protein-binding characteristics make capsules ideal for use with biologically sensitive solutions.

  • Pore size 0.8/0.2μm
  • Filtration area: 440, 930, or 1900cm2
  • Vent evacuates any air for fast filtration
  • Outlet is protected to prevent touch contamination during setup
  • Standard hose barb connections
  • Durable polypropylene filter housing can withstand maximum operating pressure of 4.1 bar (60psi)
  • Presterilized ISO-driven cleanroom manufacturing ensures product consistency
  • Stamped lot numbers ensure traceability of all critical components

Suitable for pharmaceutical aqueous solutions, tissue culture media, reagent preparation, virus suspensions, particle counting solutions, cleaning and rinsing solutions.

Compliance

Pass HIMA Challenge Test for Sterilizing Grade Filters to retain >107CFU/cm2 Brevundimonas diminuta per ASTM™ 838-83 and FDA guidelines; microbial retention correlated to 100% integrity testing during manufacturing.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Polycap TC 36 Capsule Filter, 0.8/0.2 μm, sterile, with SB inlet and outlet plus filling bell (1 pc + FB)
Material (Membrane) PES
Pore Size 0.8/0.2 μm
Sterility Sterile
Filtration Area 440 cm2
For Use With (Application) Suitable for pharmaceutical aqueous solutions, tissue culture media, reagent preparation, virus suspensions, particle counting solutions, cleaning and rinsing solutions.
Housing Material Polypropylene
Inlet Connection (Metric) 6 to 10 mm
Inlet Style Stepped Barb
Max. Pressure 4.1 bar (60 psi)
Outlet Fitting 6 to 10 mm
Outlet Style Stepped Barb
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.