Learn More
Cytiva Whatman™ Potassium Iodide Test Paper on a Reel
GSA_VA
Catalog No. 0987581 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
09-875-81 Each
1 options
Catalog No. 09-875-81 Supplier Cytiva Supplier No. 2602500A
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Combines ease-of-use with unsurpassed accuracy and consistency
This rapid determination of pH values has led to many applications in laboratories and industry
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Inexpensive
  • Convenient and portable for field use
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For detecting chlorine and other oxidizing agents
Quantity Each
Type Potassium Iodide Test Paper