Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ MD050 and MD142 Series Pressure Filtration Manifolds with Infusion Cylinders

Catalog No. p-4703923 Shop All Cytiva Products
Change view
Click to view available options
Model:
MD050/4
MD142/5/3
MD142/7/3
Material:
PTFE
Stainless Steel
Volume (Metric):
200 mL
1500 mL
2200 mL
Includes:
PTFE O-Ring, 1500 mL Infusion Cylinder for 142 mm Membrane
Silicone O-Ring/PTFE O-Ring, 200 mL Infusion Cylinder for 50 mm Membrane, Rapid Seal Connector
Silicone O-Ring/PTFE O-Ring, 2200 mL Infusion Cylinder for 142 mm Membrane
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Material Volume (Metric) Includes
09-927-716 MD050/4 Stainless Steel 200 mL Silicone O-Ring/PTFE O-Ring, 200 mL Infusion Cylinder for 50 mm Membrane, Rapid Seal Connector
09-927-721 MD142/5/3 Stainless Steel 2200 mL Silicone O-Ring/PTFE O-Ring, 2200 mL Infusion Cylinder for 142 mm Membrane
09-927-726 MD142/7/3 PTFE 1500 mL PTFE O-Ring, 1500 mL Infusion Cylinder for 142 mm Membrane
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-927-716 Supplier Cytiva Supplier No. 10450450

Cytiva Pressure Filtration Manifolds come in PTFE or stainless steel. Equipped with infusion cylinders, they are suitable for batch filtration from 20 mL, filtration of liquids/gases, ‘sterile’ filtration of serum, or clear filtration of viscous media using membranes, paper, or glass fiber discs.

  • Product Type: Pressure Filtration Manifold
  • Includes: Infusion Cylinder
  • Temperature Resistance Maximum: 200°C
TRUSTED_SUSTAINABILITY

Specifications

Disposable Reusable
Material Stainless Steel
Sterility Non-sterile
For Use With (Application) Pressure Filtration
Includes Silicone O-Ring/PTFE O-Ring, 200 mL Infusion Cylinder for 50 mm Membrane, Rapid Seal Connector
Max. Pressure 4 bar (PTFE O-Ring), 10 bar (Silicone O-Ring)
Model MD050/4
Temperature (English) Max. 392°F
Temperature (Metric) Max 200°C
Type Pressure Filtration Manifold
Volume (Metric) 200 mL
Width (English) 2.8 in.
Width (Metric) 70 mm
Height (English) 9.1 in.
Height (Metric) 230 mm
Diameter (Metric) Filter 43 mm (Pre-filter), 50 mm (Filter)
Diameter (Metric) Inside 50 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.