Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Specialized Test Paper

Catalog No. 09800979 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Application):
pH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
09-800-979 pH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-800-979 Supplier Cytiva Supplier No. 2651500
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Book, starch iodide.

  • Instant pH readings.
  • Accurate for a wide range of routine pH testing.
  • Inexpensive.
  • Convenient and portable for field use.
TRUSTED_SUSTAINABILITY

Specifications

Color Indicator Starch Iodide
Format Test Papers
For Use With (Application) pH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.