Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ UNIPLATE™ 96-Well Clear Microplates

Catalog No. PF77011651 Shop All Cytiva Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
PF77011651 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PF77011651 Supplier Cytiva Supplier No. 77011651
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

One-piece molded collection plates designed to suit a variety of applications.

  • Polystyrene
  • Withstand low temperatures (-20°C) for long-term storage
  • Flat, Round or V-shaped well bottoms
  • Conform to ANSI™/SBS (American National Standards Institute, Society of Biomolecular Screening) standards for compatibility with most microplate readers and automated plate-handling devices
  • Untreated
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Description UNIPLATE microplates
Array 8 x 12
Color Clear
Material Polystyrene
Volume (Metric) Well 650 μL
No. of Wells 96
Surface Treatment Untreated
Well Shape Square, Flat Bottom (F-well)
Quantity 50/Pk.
Length (Metric) 18.31 cm
Width (Metric) 12.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.