Learn More
Cytiva Whatman™ ML050/0/03 Stainless Steel Frit for use with the ML 050 Stainless Steel Filter Holder for In-Line Filters
Catalog No. 09927420 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
ML 050 Stainless Steel Filter Holder
Catalog No. For Use With (Equipment)
09-927-420 ML 050 Stainless Steel Filter Holder
1 options
Catalog No. 09-927-420 Supplier Cytiva Supplier No. 10464103
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
The Whatman™ ML050/0/03 Stainless Steel Frit is designed for use in in-line filters with the ML 050 Stainless Steel Filter Holder. Ideal for automated systems and high-speed batch filtration with robots, the steel build reduces contamination risk from radioactive/biohazardous materials.
  • Product Type: Frit
  • Model: ML050/0/03
  • Material: Stainless Steel
  • Includes: Ring
  • For Use With (Application): Vacuum Filtration (In-Line Filters)
  • For Use With (Equipment): Whatman™ ML 050 Stainless Steel Filter Holder
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
For Use With (Application) Vacuum Filtration (In-Line Filters)
Includes Ring
Model ML050/0/03
Type Frit
For Use With (Equipment) ML 050 Stainless Steel Filter Holder