Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Wheat 100, ultra-filtered (UF)

Catalog No. 670140
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-014-0 500 g
67-013-9 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-014-0 Supplier Gibco™ Supplier No. 670140
This item is not returnable. View return policy

Wheat 100, Ultra-Filtered (UF), is hydrolyzed wheat gluten protein that has been digested for enhanced peptide content, resulting in a low concentration of free amino acids and carbohydrates.

It is an ultra-filtered supplement and is ideal for mammalian cell culture applications.

Wheat 100, UF, contains glutamine that is associated with peptides, making it a stable source. This helps minimize the ammonia generation compared to the addition of free glutamine in cell culture media. Wheat 100, UF, can also be used to enhance growth and production in microbial applications.

Wheat 100, UF, is also part of the Gibco Peptone Preview Pak, which features our new wheat, cotton, soy, and yeast peptones. These peptones provide unique nutritional profiles compared to other cell culture supplements. The Preview Pak includes five 100 gram samples*.

*The Gibco Peptone Preview Pak is non-GMP. Larger GMP packaging sizes are available.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in cool, dry conditions in well-sealed container, 2°C to 25°C
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
Endotoxin Level 0.0 to 500.0
Format Bottle, Pail
For Use With (Application) Cell Culture and Microbial Fermentation, Tissue Culture Application
pH 5 to 7.7
Product Line Gibco
Quantity 500 g
Regulatory Status For Research Use or Further Manufacturing. Not for diagnostic use or direct administration in to humans or animals
Remarks There are no animal sourced ingredients or animal derived reagents used in the manufacture of this product.
Source Animal origin-free
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.