Learn More
Description
- Clear borosilicate glass bottles, non-graduated
- Special pour lip facilitates pouring
- Packaged in partitioned carton to help prevent scratch marks on bottles
- Hazardous communication label included for each case for content ID and safety notification
- Tamper evident seal keeps lidded bottles sealed to protect contents from accidental misuse
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Color | Clear |
| Includes | Pour lip |
| Material | Borosilicate Glass (Type I) |
| Packaging | Partitioned case |
| Quantity | 12 EA |
| Closure Color | Black |
| Closure Included | Yes |
| Height (Metric) | 231 mm |
| Product Type | Media Bottle |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
