Learn More
DWK Life Sciences Wheaton™ 47mm Filtration Assembly and Accessories
SDP
Catalog No. 14209119 Shop All DWK Life Sciences Products
Change view
Click to view available options
Type:
Base
Filtration Assembly
Funnel
Support Screen Gasket
Material:
Borosilicate Glass
Glass
PTFE
PTFE-coated Glass
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Material
14-209-119 Base Glass
14-209-121 Support Screen Gasket PTFE
14-209-122 Filtration Assembly Borosilicate Glass
14-209-124 Funnel PTFE-coated Glass
14-209-112 Filtration Assembly PTFE-coated Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-209-119 Supplier DWK Life Sciences Supplier No. 419362
Add to Cart
Add to Cart
Designed for filtering viscous or proteinaceous solutions to produce ultraclean filtrate. Wheaton™ 47mm Filtration Assembly and Accessories are manufactured from the highest quality materials.
  • Includes 300mL graduated funnel
  • Includes PTFE support screen gasket
  • Includes anodized clamp and a No. 8 silicone stopper
  • Includes a 316 stainless steel support with 120 mesh screen
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP type I, ASTM E 438 type I, Class A
For Use With (Equipment) Filtration Assembly (47 mm)
Material Glass
No. per Pack 1 Ea.
Type Base