Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ Amber Serum Vials, 10 mL, Sold by MilliporeSigma™ Supelco™

Catalog No. 111009664 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
No. per Pack:
288/Pk.
36/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
11-100-9664 288/Pk.
11-101-1397 36/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-9664 Supplier MilliporeSigma Supelco Supplier No. 33144
Only null left
Add to Cart
Add to Cart

Serum Bottles have a 20 mm, crimp top opening. Closures are not included.

TRUSTED_SUSTAINABILITY

Specifications

Description No Closure Included
Diameter (Metric) Opening 20 mm
Color Amber
Diameter (Metric) Outer 25 mm
Material Glass
Volume (Metric) 10 mL
No. per Pack 288/Pk.
Closure Type Crimp Cap
Autoclavable Autoclavable
Height (Metric) 50 mm
Product Type Serum Bottle
Show More Show Less

WHEATON is a registered trademark of Wheaton Industries, Inc.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.