Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with Metal Foil Liner
SDP

Catalog No. 0645036 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
28-400
33-400
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
06-450-36 28-400
06-450-37 33-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-450-36 Supplier DWK Life Sciences Supplier No. 240319
Add to Cart
Add to Cart

Features an aluminum foil faced pulp board liner that can be used with organic solvents

  • Good resistance to hydrocarbons, oils, ketones and alcohols
  • Not good for acids or alkalis
  • Closure size: 33mm-400
  • Phenolic
  • Autoclavable
  • Black

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 100 Cs.
Neck Finish 28-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
Closure Liner Metal Foil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.