Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with PTFE-Faced Rubber Liner
SDP

Catalog No. 0640636 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
13-425
15-415
15-425
18-400
18-415
20-400
28-400
33-400
33-430
No. per Case:
100 Cs.
200 Cs.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
06-406-36 18-400 200 Cs.
06-406-33 33-430 100 Cs.
06-450-44 15-425 200 Cs.
06-450-47 20-400 100 Cs.
06-450-49 28-400 100 Cs.
06-450-51 15-415 200 Cs.
06-450-50 33-400 100 Cs.
02-912-525 18-415 200 Cs.
06-406-40 13-425 200 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 06-406-36 Supplier DWK Life Sciences Supplier No. 240415
Add to Cart
Add to Cart

Features PTFE faced (14B) styrene-butadiene rubber liner

  • Designed for product safety
  • TFE/Styrene-Butadiene rubber liner
  • Screw cap
  • Pheolic
  • Black
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 200 Cs.
Neck Finish 18-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
Closure Liner PTFE/14B Rubber
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.