Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Caps With PE Poly-Seal Liner
SDP

Catalog No. 0645014 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
13-425
18-400
28-400
38-430
No. per Case:
100 Cs.
144 Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
06-450-14 28-400 100 Cs.
06-450-16 38-430 100 Cs.
02-912-526 18-400 144 Cs.
02-984-3 13-425 144 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-450-14 Supplier DWK Life Sciences Supplier No. 240119
Add to Cart
Add to Cart

Unique cone liner design offers superior torque retention and excellent sealing characteristics

  • Poly-Seal liner is manufactured from low density polyethylene
  • Screw Cap
  • Phenolic
  • Color: Black

It is recommended that liner be tested prior to use for leak resistant applications

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 100 Cs.
Neck Finish 28-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
Closure Liner Polyethylene
Closure Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.