Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Capillary Tubes
SDP

Catalog No. 1370747 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
1 to 5 μL
10 to 50 μL
100 to 200 μL
5 to 25 μL
Color:
Green
Red
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
13-707-47 5 to 25 μL White
13-707-46 1 to 5 μL White
13-707-48 10 to 50 μL Green
13-707-50 100 to 200 μL Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-707-47 Supplier DWK Life Sciences Supplier No. 851322
Add to Cart
Add to Cart

Glass Capillaries for Acura 841 Positive Displacement Micropipette

TRUSTED_SUSTAINABILITY

Specifications

Material Borosilicate Glass
Color White
Volume (Metric) 5 to 25 μL
Type Capillary Tube
For Use With (Equipment) Socorex Positive Displacement Micropipette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.