Description
- Inert contact surface
- Easy to assemble
- Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
- Phenolic connectors have a working temperature of up to 180°C
- Autoclavable
Specifications
Specifications
| Thread Size | 20-400, 24-400 |