Learn More
DWK Life Sciences Wheaton™ Diazomethane Generators for Gas Chromatography
SDP
Catalog No. 1420998 Shop All DWK Life Sciences Products
Change view
Click to view available options
Height (Metric):
20 cm
Catalog No. Height (Metric)
14-209-98 20 cm
1 options
Catalog No. 14-209-98 Supplier DWK Life Sciences Supplier No. 281155
Add to Cart
Add to Cart
For generating diazomethane and other gases for gas chromatography, mass sepctroscopy and NMR studies
  • Utilizes clear-seal grease-free joints
  • «O« ring not required
  • Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASMT™ E 438 Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E 438 Type I, Class A
Height (English) 7.8 in.
Height (Metric) 20 cm
Includes O-ring clamp, a 13-425 screw cap with a cut-out top and a ptfe faced rubber liner
Type Diazomethane Generator