Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Disposable 200μL Pipette Microtips
SDP

Catalog No. 1370766 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Clear
Yellow
Sterility:
Non-sterile
Sterile
Format:
Rack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Format
13-707-66 Clear Non-sterile Rack
13-707-67 Clear Sterile Rack
13-707-70 Yellow Non-sterile Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-707-66 Supplier DWK Life Sciences Supplier No. 851246

Premium pipette tips covering most research and routine applications, compatible with most pipettes

  • For both fixed- and variable-volume models
  • Precision injection molding care for air-tight fit
  • Made from high-grade, virgin polypropylene
  • Free from trace metals for excellent accuracy and reproducibility

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Disposable Single-use
For Use With (Equipment) Socorex Acura Calibra, Socorex Acura
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.