Learn More
DWK Life Sciences Disposable Syringes for Socorex 411 Step-Pett Repeater Pipette
Only three sizes cover full volume range from 10 to 5000μL. PE / PP materials
Supplier: DWK Life Sciences 851616
Description
- Supplied non-sterile in bulk 100/pk
Specifications
| Polypropylene | |
| Repeater Syringe Pipette | |
| 0.75 mL | |
| Socorex™ 411 Step-Pett™ Repeater™ pipette |
| Yellow | |
| Single-use | |
| Micro | |
| Sterile |