Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Extended Controlled 20mm LDPE Dropper Tips
SDP

Catalog No. 06450141 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100
100 Cs.
1000 Cs.
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material No. per Case
06-450-141 Orange LDPE 100 Cs.
03-440-32 Blue Polyethylene 1000 Cs.
06-450-142 Blue LDPE 100 Cs.
03-440-30 Green Polyethylene 1000 Cs.
06-450-144 Green LDPE 100 Cs.
03-440-36 Natural Polyethylene 1000 Cs.
03-440-33 Orange Polyethylene 1000 Cs.
03-440-35 Pink Polyethylene 1000 Cs.
03-440-28 Purple Polyethylene 1000 Cs.
03-440-34 Red Polyethylene 1000 Cs.
06-450-140 Red LDPE 100 Cs.
03-440-25 White Polyethylene 1000 Cs.
06-450-148 White LDPE 100 Cs.
03-440-27 Yellow Polyethylene 1000 Cs.
06-450-145 Yellow LDPE 100 Cs.
12-141-616 Natural LDPE 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 06-450-141 Supplier DWK Life Sciences Supplier No. 242433
Add to Cart
Add to Cart

20mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 100
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Material LDPE
No. per Case 100 Cs.
Certifications/Compliance ISO 9001
Drop Size 41 μL
Product Type Dropper Tip
For Use With (Equipment) Plastic dropping bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.