Learn More
Description
- Manufactured from heavy-wall borosilicate glass for durability and chemical resistance
- Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
- Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
- Acrylic rack holds up to six hybridization bottles of different sizes for safe transport or storage
- Ideal for nucleic acid and protein blotting procedures that require a minimum of probe solution
Order Info
Items come as one per case
Specifications
Specifications
| Closure Type | Screw Cap |
| Diameter (Metric) Inner | 70 mm |
| Diameter (English) Inner | 2.75 in. |
| Material | Borosilicate Glass |
| Product Type | Hybridization Bottle |
| Length (Metric) | 300 mm |
| Closure Size | 45 mm |
| Length (English) | 11.81 in. |
| For Use With (Application) | Nucleic acid and protein blotting procedures that require a minimum of probe solution |
| Liner Material | PTFE or Silicone |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
