Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips
SDP

Catalog No. 06450110 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100 Cs.
1000 Cs.
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material No. per Case
06-450-110 Red LDPE 100 Cs.
03-440-63B Blue Polyethylene 1000 Cs.
06-450-112 Blue LDPE 100 Cs.
03-440-63A Gray Polyethylene 1000 Cs.
06-450-113 Gray LDPE 100 Cs.
03-440-62 Green Polyethylene 1000 Cs.
06-450-114 Green LDPE 100 Cs.
03-440-63F Natural LDPE 1000 Cs.
03-440-63C Orange Polyethylene 1000 Cs.
03-440-63E Pink LDPE 1000 Cs.
06-450-109 Pink LDPE 100 Cs.
03-440-61 Purple Polyethylene 1000 Cs.
06-450-115 Purple LDPE 100 Cs.
03-440-63D Red Polyethylene 1000 Cs.
06-450-118 White LDPE 100 Cs.
03-440-60 Yellow Polyethylene 1000 Cs.
06-450-116 Yellow LDPE 100 Cs.
06-450-111 Orange LDPE 100 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 06-450-110 Supplier DWK Life Sciences Supplier No. 242402
Add to Cart
Add to Cart

8mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 100
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material LDPE
No. per Case 100 Cs.
Certifications/Compliance ISO 9001
Drop Size 41 μL
Product Type Dropper Tip
For Use With (Equipment) Plastic dropping bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.