Learn More
Description
For use with Snap Cap sample bottles.
- Natural low density polyethylene
- Snap Cap
- Color: Natural
- No liner
Specifications
Specifications
| Diameter (Metric) Inner | 30 mm |
| Color | Natural |
| Material | LDPE |
| No. per Case | 200/Cs. |
| For Use With (Equipment) | Snap Cap Sample Bottle |
| Certifications/Compliance | ISO 9001 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
